Gene Transcripts

Gene Symbol
Fbxw7
Entrez Gene ID
50754
Gene Name
F-box and WD-40 domain protein 7
Gene Aliases
1110001A17Rik, AGO, Cdc4, Fbw7, Fbwd6, Fbx30, Fbxo30, Fbxw6, SEL-10
Chromosome Location
Chr. 3: 84815577 - 84979198 on Build GRCm38
UniGene ID
Mm.196475
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080428.3 NP_536353.2 1 243
GenBank mRNA AB221593.1 - 1 85
AF391192.1 - 1 243
AF427101.1 - 1 165
BC131648.1 - 1 107 40110349

View Full Product Details