Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_019921.3 | NP_064305.2 | 7 | 1340 | |
GenBank mRNA | AF021833.1 | - | 4 | 694 | |
AK045234.1 | - | 1 | 2224 | ||
AK049399.1 | - | 7 | 1309 | ||
AK051044.1 | - | 5 | 975 | ||
AK083255.1 | - | 7 | 1309 | ||
AK084681.1 | - | 1 | 809 | ||
BC038483.1 | - | 4 | 398 | ||
BC054105.1 | - | 4 | 643 | 30022728 |