Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_024216.1 | NP_077178.1 | 4, 5 | 552 | |
GenBank mRNA | AK016996.1 | - | 4, 5 | 483 | |
AK042405.1 | - | 1175 | |||
AK080720.1 | - | 4, 5 | 499 | ||
AK087968.1 | - | 4, 5 | 503 | ||
AK146783.1 | - | 4, 5 | 490 | ||
BC003341.1 | - | 4, 5 | 552 | 3257412 |