Gene Transcripts

Gene Symbol
Synpo2l
Entrez Gene ID
68760
Gene Name
synaptopodin 2-like
Gene Aliases
1110054M18Rik, Chap
Chromosome Location
Chr. 14: 20658945 - 20668349 on Build GRCm38
UniGene ID
Mm.35789
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175132.3 NP_780341.1 3 830
GenBank mRNA AK053054.1 - 3 830
BC137972.1 - 3 801 8860991
BC145070.1 - 3 801

View Full Product Details