Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001111304.1 | NP_001104774.1 | 21 | 3923 | |
NM_027758.4 | NP_082034.1 | 18 | 3224 | ||
GenBank mRNA | AK017015.1 | - | 7 | 742 | |
AK049049.1 | - | 18 | 3225 | ||
AK077081.1 | - | 7 | 756 | ||
AK134614.1 | - | 21 | 3200 | ||
AK173050.1 | - | 21 | 3096 | ||
BC057027.1 | - | 17 | 2511 | 6825869 |