Gene Transcripts

Gene Symbol
Gstt4
Entrez Gene ID
75886
Gene Name
glutathione S-transferase, theta 4
Gene Aliases
4930583C14Rik, AI507011
Chromosome Location
Chr. 10: 75814943 - 75822563 on Build GRCm38
UniGene ID
Mm.268996
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029472.3 NP_083748.3 2 304
GenBank mRNA AK016340.1 - 2 288
BC052144.1 - 2 313
BC061011.1 - 2 304 6774382

View Full Product Details