Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_029857.3 | NP_084133.1 | 6 | 1056 | |
GenBank mRNA | AK014571.1 | - | 5 | 657 | |
AK029092.1 | - | 6 | 1056 | ||
AK054337.1 | - | 6 | 1053 | ||
AK148086.1 | - | 1381 | |||
AK170619.1 | - | 6 | 785 | ||
AK171189.1 | - | 6 | 802 | ||
BC013471.1 | - | 5 | 682 | 4221072 |