Gene Transcripts

Gene Symbol
Slmap
Entrez Gene ID
83997
Gene Name
sarcolemma associated protein
Gene Aliases
D330001L02Rik, Slap, mKIAA1601
Chromosome Location
Chr. 14: 26413168 - 26534624 on Build GRCm38
UniGene ID
Mm.36769
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032008.3 NP_114397.3 22 2643
GenBank mRNA AF304451.1 - 10 1191
AK129403.1 - 22 2643

View Full Product Details