Gene Transcripts

Gene Symbol
Oc90
Entrez Gene ID
18256
Gene Name
otoconin 90
Gene Aliases
Ocn-95, PLA2L, Pla2ll, oc95
Chromosome Location
Chr. 15: 65876050 - 65912411 on Build GRCm38
UniGene ID
Mm.41776
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010953.2 NP_035083.1 5 376
GenBank mRNA AF091846.1 - 3 230
AF093591.1 - 5 486
AK012981.1 - 5 378
BC048716.1 - 5 343 6334417

View Full Product Details