Gene Transcripts

Gene Symbol
Fbxw13
Entrez Gene ID
211305
Gene Name
F-box and WD-40 domain protein 13
Gene Aliases
E330017E16
Chromosome Location
Chr. 9: 109179227 - 109195975 on Build GRCm38
UniGene ID
Mm.225242
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177598.3 NP_808266.1 9 1216
GenBank mRNA AK054339.1 - 9 1217
BC119542.1 - 9 1203 40056515
BC127237.1 - 8 1039 40056507

View Full Product Details