Gene Transcripts

Gene Symbol
Itpka
Entrez Gene ID
228550
Gene Name
inositol 1,4,5-trisphosphate 3-kinase A
Gene Aliases
Chromosome Location
Chr. 2: 119742239 - 119751253 on Build GRCm38
UniGene ID
Mm.65337
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146125.2 NP_666237.1 5 1101
GenBank mRNA BC027291.1 - 5 1101

View Full Product Details