Gene Transcripts
- Gene Symbol
- Vmn2r7
- Entrez Gene ID
- 319217
- Gene Name
- vomeronasal 2, receptor 7
- Gene Aliases
- 4933425M15Rik
- Chromosome Location
- Chr. 3: 64690660 - 64719602 on Build GRCm38
- UniGene ID
- Mm.246646
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_175674.3 |
NP_783605.2
|
5 |
2155 |
|
View Full Product Details