Gene Transcripts

Gene Symbol
Skint2
Entrez Gene ID
329919
Gene Name
selection and upkeep of intraepithelial T cells 2
Gene Aliases
A430070M19, B7S3, OTTMUSG00000008540, Skint-2
Chromosome Location
Chr. 4: 112613609 - 112652248 on Build GRCm38
UniGene ID
Mm.259806
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001285963.1 NP_001272892.1 4 459
NM_001285965.1 NP_001272894.1 4 459
GenBank mRNA AK040156.1 - 3 448
BC042786.1 - 4 626 3603407
EF494892.1 - 4 459
EF494893.1 - 3 459
EU099299.1 - 4 459
EU099300.1 - 4 459
EU650234.1 - 3 433
EU650235.1 - 3 433

View Full Product Details