Gene Transcripts

Gene Symbol
Rbm20
Entrez Gene ID
73713
Gene Name
RNA binding motif protein 20
Gene Aliases
1110018J23Rik, 2010003H22Rik, AI646761
Chromosome Location
Chr. 19: 53660989 - 53867083 on Build GRCm38
UniGene ID
Mm.313695
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170847.1 NP_001164318.1 2 1362
GenBank mRNA AK142175.1 - 2 1363

View Full Product Details