Gene Transcripts

Gene Symbol
Gsdma3
Entrez Gene ID
450219
Gene Name
gasdermin A3
Gene Aliases
Bsk, Dfl, Fgn, Gsdm1l, Gsdm3, Rco2, Rim3
Chromosome Location
Chr. 11: 98626360 - 98638308 on Build GRCm38
UniGene ID
Mm.371065
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007461.1 NP_001007462.1 3 331
GenBank mRNA AY679090.1 - 3 331

View Full Product Details