Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_016880.2 | NP_058576.2 | 5, 6 | 1035 | |
GenBank mRNA | AF020790.1 | - | 5, 6 | 1036 | |
BC100542.1 | - | 5, 6 | 1035 | 30295313 | |
BC140965.1 | - | 5, 6 | 987 | 9055577 |