Gene Transcripts

Gene Symbol
Emc4
Entrez Gene ID
68032
Gene Name
ER membrane protein complex subunit 4
Gene Aliases
2610318K02Rik, Tmem85
Chromosome Location
Chr. 2: 112363019 - 112368027 on Build GRCm38
UniGene ID
Mm.220942
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026519.3 NP_080795.1 3 404
GenBank mRNA AK003668.1 - 3 372
AK012047.1 - 3 405
AK078165.1 - 3 405
AK167745.1 - 3 356
BC051926.1 - 3 360 5710720

View Full Product Details