Gene Transcripts
- Gene Symbol
- Kcnip1
- Entrez Gene ID
- 70357
- Gene Name
- Kv channel-interacting protein 1
- Gene Aliases
- KCHIP1, Kchip1.2
- Chromosome Location
- Chr. 11: 33629339 - 33993193 on Build GRCm38
- UniGene ID
- Mm.252514
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001190885.1 |
NP_001177814.1
|
8, 9 |
1030 |
|
NM_001190886.1 |
NP_001177815.1
|
7, 8 |
986 |
|
NM_001290690.1 |
NP_001277619.1
|
7, 8 |
1185 |
|
NM_027398.3 |
NP_081674.2
|
7, 8 |
997 |
|
GenBank mRNA |
AB075041.1 |
- |
7, 8 |
627 |
|
AJ278534.1 |
- |
6, 7 |
555 |
|
AK014313.1 |
- |
7, 8 |
985 |
|
AK045948.1 |
- |
8, 9 |
1031 |
|
AK046398.1 |
- |
8, 9 |
1019 |
|
AK081845.1 |
- |
7, 8 |
987 |
|
AY050525.1 |
- |
8, 9 |
635 |
|
AY050526.1 |
- |
7, 8 |
602 |
|
BC034241.1 |
- |
7, 8 |
939 |
4503251 |
DQ148493.1 |
- |
7, 8 |
939 |
|
DQ148494.1 |
- |
7, 8 |
987 |
|
DQ148495.1 |
- |
8, 9 |
635 |
|
View Full Product Details