Gene Transcripts

Gene Symbol
Slc25a18
Entrez Gene ID
71803
Gene Name
solute carrier family 25 (mitochondrial carrier), member 18
Gene Aliases
1500015I14Rik, AW125787
Chromosome Location
Chr. 6: 120773510 - 120794169 on Build GRCm38
UniGene ID
Mm.425340
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081048.2 NP_001074517.1 6 440
GenBank mRNA AK005250.1 - 4 352
BC116828.1 - 6 440 40126147
BC137603.1 - 6 440 8860622

View Full Product Details