Gene Transcripts
- Gene Symbol
- Sept9
- Entrez Gene ID
- 53860
- Gene Name
- septin 9
- Gene Aliases
- MSF1, Msf, PNUTL4, Sint1
- Chromosome Location
- Chr. 11: 117199661 - 117362325 on Build GRCm38
- UniGene ID
- Mm.38450
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC094455.1 |
- |
|
2866 |
|
- Gene Symbol
- Clic1
- Entrez Gene ID
- 114584
- Gene Name
- chloride intracellular channel 1
- Gene Aliases
- Clcp, G6
- Chromosome Location
- Chr. 17: 35050243 - 35058719 on Build GRCm38
- UniGene ID
- Mm.29524
- Species
- Mus musculus
View Full Product Details