Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_029512.2 | NP_083788.2 | 5 | 1134 | |
NM_181734.3 | NP_859423.2 | 5 | 1131 | ||
GenBank mRNA | AK018052.1 | - | 4 | 900 | |
AK033600.1 | - | 5 | 1080 | ||
AK039450.1 | - | 5 | 1132 | ||
AK043791.1 | - | 5 | 1101 | ||
AK081680.1 | - | 5 | 1094 | ||
AK156641.1 | - | 5 | 1097 | ||
BC052389.1 | - | 5 | 1090 | 5718874 | |
BC060726.1 | - | 4 | 1000 |