Gene Transcripts

Gene Symbol
Ttll3
Entrez Gene ID
101100
Gene Name
tubulin tyrosine ligase-like family, member 3
Gene Aliases
4833441J24Rik, AI450050
Chromosome Location
Chr. 6: 113389260 - 113414583 on Build GRCm38
UniGene ID
Mm.388573
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133923.6 NP_598684.4 11 1978
GenBank mRNA AK080321.1 - 7 1392
AM690746.1 - 11 1978
BC006830.1 - 8 1172

View Full Product Details