Gene Transcripts

Gene Symbol
Cobl
Entrez Gene ID
12808
Gene Name
cordon-bleu WH2 repeat
Gene Aliases
Chromosome Location
Chr. 11: 12236676 - 12464997 on Build GRCm38
UniGene ID
Mm.22847
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282993.1 NP_001269922.1 4 775
NM_001282994.1 NP_001269923.1 4 606
NM_172496.3 NP_766084.3 4 775
GenBank mRNA AK028833.1 - 4 622
AK049730.1 - 4 568
AK122334.1 - 4 606
AK147366.1 - 4 776
AK147633.1 - 4 776
AY308745.1 - 4 615
BC145116.1 - 4 615
BC145118.1 - 4 615 9053666
BC150706.1 - 4 615 9087617
U26967.1 - 3 348

View Full Product Details