Gene Transcripts

Gene Symbol
Dbil5
Entrez Gene ID
13168
Gene Name
diazepam binding inhibitor-like 5
Gene Aliases
ELP
Chromosome Location
Chr. 11: 76217613 - 76218665 on Build GRCm38
UniGene ID
Mm.347413
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021294.2 NP_067269.1 2 258
GenBank mRNA AK006528.1 - 2 219
BC048474.1 - 2 237 6773240
CN833562.1 - 261
S83465.1 - 2 245

View Full Product Details