Gene Transcripts
- Gene Symbol
- Psma3
- Entrez Gene ID
- 19167
- Gene Name
- proteasome (prosome, macropain) subunit, alpha type 3
- Gene Aliases
- Lmpc8
- Chromosome Location
- Chr. 12: 70974623 - 70994937 on Build GRCm38
- UniGene ID
- Mm.296338
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_011184.4 |
NP_035314.3
|
1, 2 |
143 |
|
GenBank mRNA |
AF055983.1 |
- |
1, 2 |
61 |
|
AF060089.1 |
- |
1, 2 |
29 |
|
AI303320.1 |
- |
1, 2 |
142 |
|
AK002877.1 |
- |
1, 2 |
74 |
|
AK165424.1 |
- |
1, 2 |
96 |
|
AK168495.1 |
- |
1, 2 |
48 |
|
AK169577.1 |
- |
1, 2 |
46 |
|
AK171669.1 |
- |
1, 2 |
52 |
|
AK172369.1 |
- |
1, 2 |
52 |
|
BC091743.1 |
- |
1, 2 |
33 |
5030475 |
View Full Product Details