Gene Transcripts
- Gene Symbol
- Trove2
- Entrez Gene ID
- 20822
- Gene Name
- TROVE domain family, member 2
- Gene Aliases
- 1810007I17Rik, A530054J02Rik, AI646302, SS-A/Ro, Ssa, Ssa2
- Chromosome Location
- Chr. 1: 143750790 - 143777051 on Build GRCm38
- UniGene ID
- Mm.40370
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_013835.2 |
NP_038863.1
|
2, 3 |
742 |
|
GenBank mRNA |
AK076167.1 |
- |
2, 3 |
719 |
|
AK167379.1 |
- |
2, 3 |
683 |
|
AK167555.1 |
- |
2, 3 |
683 |
|
AK167645.1 |
- |
2, 3 |
683 |
|
BC040368.1 |
- |
1, 2 |
613 |
|
BC051974.1 |
- |
1, 2 |
619 |
5697399 |
BC052380.1 |
- |
1, 2 |
619 |
5697399 |
L81154.1 |
- |
1, 2 |
590 |
|
U66843.1 |
- |
2, 3 |
669 |
|
View Full Product Details