Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_027748.3 | NP_082024.2 | 3 | 954 | |
GenBank mRNA | AJ292189.1 | - | 3 | 745 | |
AK017128.1 | - | 1 | 1048 | ||
AK030878.1 | - | 1 | 23 | ||
AK033848.1 | - | 3 | 604 | ||
AK051249.1 | - | 1 | 325 | ||
AK154372.1 | - | 3 | 963 | ||
BC039989.1 | - | 3 | 910 | ||
BC089030.1 | - | 3 | 793 | ||
BC094546.1 | - | 3 | 911 | ||
BC137617.1 | - | 3 | 895 | 8860636 | |
BC137618.1 | - | 3 | 895 | 8860637 |