Gene Transcripts

Gene Symbol
Taar2
Entrez Gene ID
209512
Gene Name
trace amine-associated receptor 2
Gene Aliases
Gm224, Gm225, Gpr58, Tarr2
Chromosome Location
Chr. 10: 23938525 - 23960552 on Build GRCm38
UniGene ID
Mm.300611
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007266.1 NP_001007267.1 2 979
GenBank mRNA AY702326.1 - 2 979
BC139118.1 - 2 1032 8862140
BC145814.1 - 2 1032 40131117

View Full Product Details