Gene Transcripts

Gene Symbol
Ado
Entrez Gene ID
211488
Gene Name
2-aminoethanethiol (cysteamine) dioxygenase
Gene Aliases
Gm237
Chromosome Location
Chr. 10: 67544511 - 67548955 on Build GRCm38
UniGene ID
Mm.297250
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005419.2 NP_001005419.2 1 503
GenBank mRNA AK155334.1 - 1 506
BC057106.1 - 1 490 5721417
BC058407.1 - 1 490 5721417

View Full Product Details