Gene Transcripts

Gene Symbol
Crygn
Entrez Gene ID
214301
Gene Name
crystallin, gamma N
Gene Aliases
Chromosome Location
Chr. 5: 24751002 - 24757843 on Build GRCm38
UniGene ID
Mm.63484
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153076.2 NP_694716.2 2 156
GenBank mRNA AF445456.1 - 2 156
BC120913.1 - 2 156 40091405
BC120914.1 - 1 145
CF739670.1 - 1 140

View Full Product Details