Gene Transcripts

Gene Symbol
Csnka2ip
Entrez Gene ID
224291
Gene Name
casein kinase 2, alpha prime interacting protein
Gene Aliases
Ckt2
Chromosome Location
Chr. 16: 64477810 - 64602366 on Build GRCm38
UniGene ID
Mm.246580
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173861.2 NP_776286.1 1 510
GenBank mRNA AF463503.1 - 1 553
BC137818.1 - 1 432 8860837

View Full Product Details