Gene Transcripts

Gene Symbol
Fam160b2
Entrez Gene ID
239170
Gene Name
family with sequence similarity 160, member B2
Gene Aliases
G430067P06Rik, Rai16
Chromosome Location
Chr. 14: 70583293 - 70599835 on Build GRCm38
UniGene ID
Mm.247582
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_194345.1 NP_919326.1 12, 13 1662
GenBank mRNA AK090035.1 - 7, 8 913
CN529793.1 - 3, 4 364

View Full Product Details