Gene Transcripts

Gene Symbol
Krtap24-1
Entrez Gene ID
239932
Gene Name
keratin associated protein 24-1
Gene Aliases
Gm312
Chromosome Location
Chr. 16: 88610709 - 88612279 on Build GRCm38
UniGene ID
Mm.120273
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163141.1 NP_001156613.1 1 478
GenBank mRNA AK037168.1 - 1 479

View Full Product Details