Gene Transcripts

Gene Symbol
Megf8
Entrez Gene ID
269878
Gene Name
multiple EGF-like-domains 8
Gene Aliases
AW049492, Egfl4, b2b1702Clo, b2b288Clo, m687Ddg, mKIAA0817
Chromosome Location
Chr. 7: 25316742 - 25365917 on Build GRCm38
UniGene ID
Mm.38527
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001160400.1 NP_001153872.1 10 2025
GenBank mRNA EU723517.1 - 10 2025

View Full Product Details