Gene Transcripts

Gene Symbol
Vmn2r23
Entrez Gene ID
435916
Gene Name
vomeronasal 2, receptor 23
Gene Aliases
EG435916
Chromosome Location
Chr. 6: 123702821 - 123742239 on Build GRCm38
UniGene ID
Mm.469874
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001104638.1 NP_001098108.1 4 1342

View Full Product Details