Gene Transcripts

Gene Symbol
Cts6
Entrez Gene ID
58518
Gene Name
cathepsin 6
Gene Aliases
1600022N02Rik
Chromosome Location
Chr. 13: 61195147 - 61203392 on Build GRCm38
UniGene ID
Mm.143794
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021445.1 NP_067420.1 4 400
GenBank mRNA AF223401.1 - 4 400
AK005518.1 - 4 419
AY014781.1 - 4 416
BC099455.1 - 4 402 30787134

View Full Product Details