Gene Transcripts

Gene Symbol
Mrps16
Entrez Gene ID
66242
Gene Name
mitochondrial ribosomal protein S16
Gene Aliases
1500011E11Rik, Mprs16
Chromosome Location
Chr. 14: 20391231 - 20393555 on Build GRCm38
UniGene ID
Mm.23747
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025440.2 NP_079716.1 3 438
GenBank mRNA AB049949.1 - 3 496
AK005206.1 - 3 439
AK019243.1 - 3 384
BC024656.1 - 3 404 3986314

View Full Product Details