Gene Transcripts

Gene Symbol
Emc2
Entrez Gene ID
66736
Gene Name
ER membrane protein complex subunit 2
Gene Aliases
4921531G14Rik, AV060620, AW209495, Ttc35
Chromosome Location
Chr. 15: 43477214 - 43527777 on Build GRCm38
UniGene ID
Mm.244512
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025736.2 NP_080012.1 9, 10 757
GenBank mRNA AF332065.1 - 9, 10 685
AK014989.1 - 9, 10 758
AK015968.1 - 9, 10 723

View Full Product Details