Gene Transcripts

Gene Symbol
Fcrl6
Entrez Gene ID
677296
Gene Name
Fc receptor-like 6
Gene Aliases
ENSMUSG00000070504, FcRH6, mIFGP6, moFcRH6
Chromosome Location
Chr. 1: 172596640 - 172618322 on Build GRCm38
UniGene ID
Mm.439612
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164725.1 NP_001158197.1 3 193
GenBank mRNA DQ470838.1 - 3 198
DQ470839.1 - 3 198
DQ470840.1 - 3 189
EF032497.1 - 3 102

View Full Product Details