Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001146342.2 | NP_001139814.2 | 4 | 833 | |
NM_001167818.1 | NP_001161290.1 | 6 | 1082 | ||
GenBank mRNA | AK018322.1 | - | 6 | 1050 | |
AK170321.1 | - | 4 | 834 | ||
BC052355.1 | - | 6 | 913 | 6770333 | |
DQ788834.1 | - | 6 | 859 |