Gene Transcripts

Gene Symbol
Rnaseh2c
Entrez Gene ID
68209
Gene Name
ribonuclease H2, subunit C
Gene Aliases
1500026D16Rik, AYP1
Chromosome Location
Chr. 19: 5601873 - 5602959 on Build GRCm38
UniGene ID
Mm.246200
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026616.2 NP_080892.1 2 278
GenBank mRNA AF346604.1 - 2 276
AK011556.1 - 2 256
AK018776.1 - 2 267
BC024333.1 - 2 250 5040687

View Full Product Details