Gene Transcripts

Gene Symbol
Tmco2
Entrez Gene ID
69469
Gene Name
transmembrane and coiled-coil domains 2
Gene Aliases
1700028F16Rik
Chromosome Location
Chr. 4: 121105651 - 121109226 on Build GRCm38
UniGene ID
Mm.481120
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081312.1 NP_001074781.1 2 405
GenBank mRNA AK006456.1 - 2 406

View Full Product Details