Gene Transcripts

Gene Symbol
Izumo4
Entrez Gene ID
71564
Gene Name
IZUMO family member 4
Gene Aliases
9030607L17Rik, AI550407
Chromosome Location
Chr. 10: 80702199 - 80705382 on Build GRCm38
UniGene ID
Mm.35802
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027829.3 NP_082105.3 1, 2 276
GenBank mRNA BC057953.1 - 1, 2 202
BF682661.1 - 1, 2 211
CF105977.1 - 1, 2 276

View Full Product Details