Gene Transcripts

Gene Symbol
Cib4
Entrez Gene ID
73259
Gene Name
calcium and integrin binding family member 4
Gene Aliases
1700041E20Rik
Chromosome Location
Chr. 5: 30485583 - 30545837 on Build GRCm38
UniGene ID
Mm.443299
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028483.1 NP_082759.1 3 220
GenBank mRNA AK006670.1 - 3 221
BC099558.1 - 3 166
BC118938.1 - 3 185 40091772

View Full Product Details