Gene Transcripts

Gene Symbol
Rhox13
Entrez Gene ID
73614
Gene Name
reproductive homeobox 13
Gene Aliases
1700123J19Rik
Chromosome Location
Chr. X: 38120840 - 38129966 on Build GRCm38
UniGene ID
Mm.50505
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001185002.1 NP_001171931.1 1 502
GenBank mRNA AV490373.1 - 1 123

View Full Product Details