Product Details
- SNP ID
-
rs13483770
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:59235498 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTTTAATCAACTCTGGTAAAGAAA[A/G]AATGGCCTGAACAGAAACTTTCCAA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Fgf13
PubMed Links
Gene Details
- Gene
- Fgf13
- Gene Name
- fibroblast growth factor 13
There are no transcripts associated with this gene.
View Full Product Details