Product Details

SNP ID
rs6297639
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.12:41094798 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CCTGTGAGCAGGGACAGGTCAGTAG[A/T]TCTGTCTGCTCAGTGGGGAGTGACT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
Immp2l PubMed Links

Gene Details

Gene
Immp2l
Gene Name
IMP2 inner mitochondrial membrane peptidase-like (S. cerevisiae)
There are no transcripts associated with this gene.

View Full Product Details