Product Details

SNP ID
rs4214929
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.16:88037247 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GAGTTAAAAGTTTCCAGATCTCAGG[C/T]GTGAGAAGGGCAGTGACTTTGCACC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Grik1 PubMed Links

Gene Details

Gene
Grik1
Gene Name
glutamate receptor, ionotropic, kainate 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_010348.3 Intron NP_034478.1
NM_146072.4 Intron NP_666184.2

View Full Product Details