Product Details

SNP ID
rs13470759
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.2:180036944 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CACCCAAAAGTAGGCTTACCTTGAG[C/G]GGCTGATCCCGCCTCATGACGGCAA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
Lsm14b PubMed Links

Gene Details

Gene
Lsm14b
Gene Name
LSM14 homolog B (SCD6, S. cerevisiae)
There are no transcripts associated with this gene.

Gene
Psma7
Gene Name
proteasome (prosome, macropain) subunit, alpha type 7
There are no transcripts associated with this gene.

Gene
Ss18l1
Gene Name
synovial sarcoma translocation gene on chromosome 18-like 1
There are no transcripts associated with this gene.

View Full Product Details