Product Details
- SNP ID
-
rs13481276
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:3779694 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCCCCTTCCTCTACCCTTCCTCCC[A/G]TCACAGATCCCAAGCCAGAGCAAAC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Dtnb
PubMed Links
Gene Details
- Gene
- Dtnb
- Gene Name
- dystrobrevin, beta
View Full Product Details